COMH04236F70 Combi-Blocks, Inc.

4-[1-(Cyclopropylmethyl)-1h-imidazol-2-yl]piperidine dihydrochloride

  • MDL number MFCD27996002
  • Cas Number : 1823872-18-9
  • Molecular Weight : 278.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9673
Availability details for
Quantity
  • Availability
  • Add to Cart