COMH04236F74 Combi-Blocks, Inc.

(2-(2-[4-(Trifluoromethyl)phenyl]-1,3-thiazol-4-yl)ethyl)amine dihydrochloride

  • MDL number MFCD11983595
  • Cas Number : 1332531-22-2
  • Molecular Weight : 345.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9677
Availability details for
Quantity
  • Availability
  • Add to Cart