COMH04236F9F Combi-Blocks, Inc.

6-Bromo-2-(2-isopropoxyphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD03421115
  • Cas Number : 1160253-11-1
  • Molecular Weight : 404.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9720
Availability details for
Quantity
  • Availability
  • Add to Cart