COMH04236FA2 Combi-Blocks, Inc.

N-1,3-Benzodioxol-5-yl-2-cyano-3-(dimethylamino)acrylamide

  • MDL number MFCD27756638
  • Cas Number : 1786462-99-4
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9723
Availability details for
Quantity
  • Availability
  • Add to Cart