COMH04236FA6 Combi-Blocks, Inc.

8-Chloro-2-(2,4-dimethylphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD03422875
  • Cas Number : 1160256-46-1
  • Molecular Weight : 330.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9727
Availability details for
Quantity
  • Availability
  • Add to Cart