COMH04236FF2 Combi-Blocks, Inc.

4-[2-(4-Aminophenyl)ethyl]phenol

  • MDL number MFCD08458839
  • Cas Number : 33384-05-3
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9803
Availability details for
Quantity
  • Availability
  • Add to Cart