COMH0423703E Combi-Blocks, Inc.

Ethyl (6-hydroxy-2-methylpyrimidin-4-yl)acetate

  • MDL number MFCD03411877
  • Cas Number : 54554-50-6
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9879
Availability details for
Quantity
  • Availability
  • Add to Cart