COMH0423704C Combi-Blocks, Inc.

[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]amine hydrochloride

  • MDL number MFCD09759066
  • Cas Number : 132451-28-6
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9893
Availability details for
Quantity
  • Availability
  • Add to Cart