COMH0423707B Combi-Blocks, Inc.

3-[((5-[(Cyclopropylamino)carbonyl]-1,2,4-oxadiazol-3-yl)methyl)thio]propanoic acid

  • MDL number MFCD12026887
  • Cas Number : 1119449-70-5
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9940
Availability details for
Quantity
  • Availability
  • Add to Cart