COMH0423709A Combi-Blocks, Inc.

2-Mercapto-3-(1-naphthyl)quinazolin-4(3h)-one

  • MDL number MFCD01134714
  • Cas Number : 22453-82-3
  • Molecular Weight : 304.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9971
Availability details for
Quantity
  • Availability
  • Add to Cart