COMH042370B1 Combi-Blocks, Inc.

N,N-Dimethyl-6-piperidin-4-ylnicotinamide dihydrochloride

  • MDL number MFCD29035030
  • Cas Number : 1847443-82-6
  • Molecular Weight : 306.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9994
Availability details for
Quantity
  • Availability
  • Add to Cart