COMH0423710D Combi-Blocks, Inc.

2-(4-Isopropoxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03074952
  • Cas Number : 438226-06-3
  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0088
Availability details for
Quantity
  • Availability
  • Add to Cart