COMH04237112 Combi-Blocks, Inc.

4-(2-Methoxyphenyl)-1,4-dihydro[1,3,5]triazino[1,2-a]benzimidazol-2-amine

  • MDL number MFCD04159896
  • Cas Number : 669718-22-3
  • Molecular Weight : 293.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0093
Availability details for
Quantity
  • Availability
  • Add to Cart