COMH0423711E Combi-Blocks, Inc.

N-[(3-Methoxy-2-naphthyl)methyl]aniline

  • MDL number MFCD05860811
  • Cas Number : 727675-30-1
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0105
Availability details for
Quantity
  • Availability
  • Add to Cart