COMH0423713B Combi-Blocks, Inc.

2-(3-Isopropoxyphenyl)quinoline-4-carbohydrazide

  • MDL number MFCD02608591
  • Cas Number : 524733-45-7
  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0134
Availability details for
Quantity
  • Availability
  • Add to Cart