COMH0423713E Combi-Blocks, Inc.

2-(Methylamino)-n-(propan-2-yl)acetamide hydrochloride

  • MDL number MFCD13196084
  • Cas Number : 51489-14-6
  • Molecular Weight : 166.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0137
Availability details for
Quantity
  • Availability
  • Add to Cart