COMH04237142 Combi-Blocks, Inc.

2-(Azepan-1-ylsulfonyl)ethanamine

  • MDL number MFCD09041181
  • Cas Number : 926211-37-2
  • Molecular Weight : 206.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0141
Availability details for
Quantity
  • Availability
  • Add to Cart