COMH04237143 Combi-Blocks, Inc.

N-[2-(2-Fluorophenyl)ethyl]-4-oxopentanamide

  • MDL number MFCD02625641
  • Cas Number : 361464-87-1
  • Molecular Weight : 237.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0142
Availability details for
Quantity
  • Availability
  • Add to Cart