COMH04237154 Combi-Blocks, Inc.

4-Pyridin-4-ylpyrimidine-2(1h)-thione

  • MDL number MFCD11558232
  • Cas Number : 874779-72-3
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0159
Availability details for
Quantity
  • Availability
  • Add to Cart