COMH04237182 Combi-Blocks, Inc.

Propyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta[b]thiophene-3-carboxylate

  • MDL number MFCD01630698
  • Cas Number : 351982-19-9
  • Molecular Weight : 253.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0206
Availability details for
Quantity
  • Availability
  • Add to Cart