COMH0423721D Combi-Blocks, Inc.

4-(2-Phenoxyethyl)piperidine hydrochloride

  • MDL number MFCD09879252
  • Cas Number : 347873-67-0
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0363
Availability details for
Quantity
  • Availability
  • Add to Cart