COMH04237226 Combi-Blocks, Inc.

6-tert-Butyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide

  • MDL number MFCD09971927
  • Cas Number : 956576-54-8
  • Molecular Weight : 252.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0372
Availability details for
Quantity
  • Availability
  • Add to Cart