COMH0423722C Combi-Blocks, Inc.

2-(4-Sec-butylphenyl)-6-chloroquinoline-4-carboxylic acid

  • MDL number MFCD03420109
  • Cas Number : 932928-94-4
  • Molecular Weight : 339.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0378
Availability details for
Quantity
  • Availability
  • Add to Cart