COMH0423722F Combi-Blocks, Inc.

N-(4-Acetylphenyl)-4-(2,4-dichlorophenoxy)butanamide

  • MDL number MFCD00705625
  • Cas Number : 348163-50-8
  • Molecular Weight : 366.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0381
Availability details for
Quantity
  • Availability
  • Add to Cart