COMH0423725E Combi-Blocks, Inc.

1-(4-Azepan-1-yl-3-fluorophenyl)ethanone

  • MDL number MFCD02633409
  • Cas Number : 420826-76-2
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0428
Availability details for
Quantity
  • Availability
  • Add to Cart