COMH04237265 Combi-Blocks, Inc.

Ethyl 2-amino-5,6,7,8,9,10-hexahydro-4h-cyclonona[b]thiophene-3-carboxylate

  • MDL number MFCD03075348
  • Cas Number : 438230-65-0
  • Molecular Weight : 267.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0435
Availability details for
Quantity
  • Availability
  • Add to Cart