COMH0423728A Combi-Blocks, Inc.

Ethyl 4-(2-cyano-4-nitrophenyl)piperazine-1-carboxylate

  • MDL number MFCD07837383
  • Cas Number : 849484-94-2
  • Molecular Weight : 304.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0472
Availability details for
Quantity
  • Availability
  • Add to Cart