COMH0423729A Combi-Blocks, Inc.

3-Chloro-n-[3-(hydrazinocarbonyl)phenyl]benzamide

  • MDL number MFCD02255613
  • Cas Number : 911039-80-0
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0488
Availability details for
Quantity
  • Availability
  • Add to Cart