COMH0423729D Combi-Blocks, Inc.

1-[4-(1,1-Dioxido-1,2-benzisothiazol-3-yl)piperazin-1-yl]propan-2-ol

  • MDL number MFCD01579587
  • Cas Number : 346448-88-2
  • Molecular Weight : 309.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0491
Availability details for
Quantity
  • Availability
  • Add to Cart