COMH042372AF Combi-Blocks, Inc.

Methyl n-(2-ethoxyphenyl)-n-(phenylsulfonyl)glycinate

  • MDL number MFCD01964853
  • Cas Number : 358378-00-4
  • Molecular Weight : 349.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0509
Availability details for
Quantity
  • Availability
  • Add to Cart