COMH042372B5 Combi-Blocks, Inc.

N-[2-(3-Ethyl-5-methylphenoxy)ethyl]-2-hydroxybenzamide

  • MDL number MFCD08142738
  • Cas Number : 893778-77-3
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0515
Availability details for
Quantity
  • Availability
  • Add to Cart