COMH042372E2 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)quinoline-4-carbohydrazide

  • MDL number MFCD01993672
  • Cas Number : 51842-72-9
  • Molecular Weight : 293.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0561
Availability details for
Quantity
  • Availability
  • Add to Cart