COMH0423730F Combi-Blocks, Inc.

N-Cyclopropyl-4-(1h-imidazol-1-yl)-3-nitrobenzamide

  • MDL number MFCD07800550
  • Cas Number : 893769-73-8
  • Molecular Weight : 272.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0606
Availability details for
Quantity
  • Availability
  • Add to Cart