COMH04237313 Combi-Blocks, Inc.

4-[(4-([2-(4-Nitrophenyl)-2-oxoethoxy]carbonyl)phenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD06756122
  • Cas Number : 774557-89-0
  • Molecular Weight : 400.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0610
Availability details for
Quantity
  • Availability
  • Add to Cart