COMH0423731B Combi-Blocks, Inc.

3-(Benzylsulfonyl)-5-nitroaniline

  • MDL number MFCD12027074
  • Cas Number : 912772-83-9
  • Molecular Weight : 292.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0618
Availability details for
Quantity
  • Availability
  • Add to Cart