COMH04237346 Combi-Blocks, Inc.

1-(Chloroacetyl)-2-methyl-5-nitroindoline

  • MDL number MFCD13705155
  • Cas Number : 887572-47-6
  • Molecular Weight : 254.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0661
Availability details for
Quantity
  • Availability
  • Add to Cart