COMH04237397 Combi-Blocks, Inc.

(6-Methoxy-1,3-benzodioxol-5-yl)amine hydrochloride

  • MDL number MFCD30723497
  • Cas Number : 69151-32-2
  • Molecular Weight : 203.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0742
Availability details for
Quantity
  • Availability
  • Add to Cart