COMH042373A5 Combi-Blocks, Inc.

(2,4-Dimethylphenyl)(phenyl)acetonitrile

  • MDL number MFCD24849131
  • Cas Number : 82954-09-4
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0756
Availability details for
Quantity
  • Availability
  • Add to Cart