COMH042373AC Combi-Blocks, Inc.

N4-Benzyl-6-chloropyrimidine-2,4-diamine

  • MDL number MFCD00664674
  • Cas Number : 91066-67-0
  • Molecular Weight : 234.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0763
Availability details for
Quantity
  • Availability
  • Add to Cart