COMH042373B5 Combi-Blocks, Inc.

(2E)-4-[(3-Acetylphenyl)amino]-4-oxobut-2-enoic acid

  • MDL number MFCD01055665
  • Cas Number : 95695-48-0
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0772
Availability details for
Quantity
  • Availability
  • Add to Cart