COMH042373BC Combi-Blocks, Inc.

1-(3,5-Dimethyl-4-nitrophenyl)pyrrolidin-2-one

  • MDL number MFCD03196988
  • Cas Number : 869947-97-7
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0779
Availability details for
Quantity
  • Availability
  • Add to Cart