COMH042373E4 Combi-Blocks, Inc.

8-Methoxy-2-methyl-5-nitroquinoline

  • MDL number MFCD06662295
  • Cas Number : 857495-64-8
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0820
Availability details for
Quantity
  • Availability
  • Add to Cart