COMH042373EC Combi-Blocks, Inc.

5-Chloro-3-(2-methoxy-5-nitrobenzyl)-1,3-benzoxazol-2(3h)-one

  • MDL number MFCD08290355
  • Cas Number : 898796-77-5
  • Molecular Weight : 334.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0828
Availability details for
Quantity
  • Availability
  • Add to Cart