COMH0423740F Combi-Blocks, Inc.

N(1),N(1)-Dimethyl-1-[2-(trifluoromethyl)phenyl]ethane-1,2-diamine

  • MDL number MFCD09735718
  • Cas Number : 933684-53-8
  • Molecular Weight : 232.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0863
Availability details for
Quantity
  • Availability
  • Add to Cart