COMH04237417 Combi-Blocks, Inc.

N-(2-Bromo-4,6-difluorophenyl)-2-chloroacetamide

  • MDL number MFCD04127864
  • Cas Number : 737780-11-9
  • Molecular Weight : 284.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0871
Availability details for
Quantity
  • Availability
  • Add to Cart