COMH04237419 Combi-Blocks, Inc.

2-Bromo-n-(4-propoxybenzyl)acetamide

  • MDL number MFCD16488532
  • Cas Number : 1225740-34-0
  • Molecular Weight : 286.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0873
Availability details for
Quantity
  • Availability
  • Add to Cart