COMH04237439 Combi-Blocks, Inc.

Methyl n-(2-chlorophenyl)-n-(phenylsulfonyl)glycinate

  • MDL number MFCD01964849
  • Cas Number : 359770-42-6
  • Molecular Weight : 339.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0906
Availability details for
Quantity
  • Availability
  • Add to Cart