COMH0423745D Combi-Blocks, Inc.

2-(Methylamino)-n-propylacetamide

  • MDL number MFCD05270871
  • Cas Number : 901273-19-6
  • Molecular Weight : 130.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0942
Availability details for
Quantity
  • Availability
  • Add to Cart