COMH04237473 Combi-Blocks, Inc.

5-(5-Isopropylthien-3-yl)-4-propyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD03423430
  • Cas Number : 861227-42-1
  • Molecular Weight : 267.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0964
Availability details for
Quantity
  • Availability
  • Add to Cart