COMH0423749E Combi-Blocks, Inc.

1-[(4-Methylphenyl)sulfonyl]-1,4-diazepane hydrochloride

  • MDL number MFCD27756445
  • Cas Number : 872103-28-1
  • Molecular Weight : 290.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1008
Availability details for
Quantity
  • Availability
  • Add to Cart